Läs mer
MedChemExpress PAR-4 Agonist Peptide, amide TFA
Produktkod. 16439538
Encompass_Preferred
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
16439538 10 mg
1 options
Produktkod. 16439538 Leverantör MedChemExpress Leverantörsnummer HYP1309A
MedChemExpress PAR-4 Agonist Peptide, amide TFA (PAR-4-AP TFA; AY-NH2 TFA) is a proteinase-activated receptor-4 (PAR-4) agonist, which has no effect on either PAR-1 or PAR-2 and whose effects are blocked by a PAR-4 antagonist.
Specifikationer
Renhet 99.93%
CAS 1228078-65-6
Innehåll och lagring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
För användning med (applikation) COVID-19-immunoregulation
Molekylvikt (g/mol) 794.82 g/mol
Produkttyp Peptides
Kvantitet 10 mg
Frakt skick Room temperature
Löslighet DMSO : ≥ 250 mg/mL (314.54 mM) ∣H2O : 100 mg/mL (125.81 mM∣Need ultrasonic)
Synonym PAR-4-AP TFA∣AY-NH2 TFA