Läs mer
Para-Naphthol Duloxetine, TRC
Produktkod. 30135812
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135812 50 mg
30133048 250 mg
30010696 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135812 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP19186050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 949095-98-1
Molekylformel C18 H19 N O S
Molekylvikt (g/mol) 297.4146
Synonym 1-Naphthalenol, 4-[3-(methylamino)-1-(2-thienyl)propyl]-, 4-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol, Para-Naphthol duloxetine, 4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Duloxetine Hydrochloride Imp. C (EP)
IUPAC-namn 4-[3-(methylamino)-1-thiophen-2-ylpropyl]naphthalen-1-ol
LEDER CNCCC(c1cccs1)c2ccc(O)c3ccccc23
Specifikationer
Kemiskt namn eller material 4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
Kvantitet 50 mg
InChI formel InChI=1S/C18H19NOS/c1-19-11-10-16(18-7-4-12-21-18)14-8-9-17(20)15-6-3-2-5-13(14)15/h2-9,12,16,19-20H,10-11H2,1H3
Formel vikt 297.1187