Läs mer
para-Topolin, TRC
Produktkod. 30452147
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452147 1 mg
30452080 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5403001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80054-30-4
Molekylformel C12 H11 N5 O
Molekylvikt (g/mol) 241.25
Synonym 6-(4-Hydroxybenzylamino)purine, p-Topolin, 4-[(9H-Purin-6-ylamino)methyl]phenol, 6-(4-Hydroxybenzylamino)purine, 6-p-Hydroxybenzylaminopurine, 4-[(1H-purin-6-ylamino)methyl]-phenol
IUPAC-namn 4-[(7H-purin-6-ylamino)methyl]phenol
LEDER Oc1ccc(CNc2ncnc3nc[nH]c23)cc1
Specifikationer
Kemiskt namn eller material para-Topolin
Kvantitet 1 mg
InChI formel InChI=1S/C12H11N5O/c18-9-3-1-8(2-4-9)5-13-11-10-12(15-6-14-10)17-7-16-11/h1-4,6-7,18H,5H2,(H2,13,14,15,16,17)
Formel vikt 241.0964