Learn More
R&D Systems™ Parameter Immunoassay Control Set 1165 for cAMP
Product Code. 18422580
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
Product Code. Quantity Unit Size
18422580 1 Each 1 set
1 options
Product Code. 18422580 Supplier R&D Systems™ Supplier No. QC52
  • ONLINE EXCLUSIVE PRICE
    2282.89 SEK
    2013.90 SEK / 1 uppsättningar
    Save 268.99 SEK
    11% Av
 Restricted Item
Beräknad leverans: 03-12-2026
For use with catalog numbers KGE002B, SKGE002B, PKGE002B
Application: For Research Use Only
  • Contains ProClin 300 (<0.01%)
  • Suitable for research applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes cAMP
Type Immunoassay Control
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Multi-species