Läs mer
Paramethasone Acetate, TRC
Produktkod. 30135373
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135373 5 mg
17944235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135373 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1918455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1597-82-6
Molekylformel C24 H31 F O6
Molekylvikt (g/mol) 434.5
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-6-fluoro-11,17-dihydroxy-16-methyl-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-acetate (6CI,7CI,8CI), 16α-Methyl-6α-fluoroprednisolone 21-acetate, 21-Acetoxy-6α-fluoro-11β,17-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, 21-Acetyl-6α-fluoro-16α-methylprednisolone, 6α-Fluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-acetate, 6α-Fluoro-16α-methylprednisolone 21-acetate, Alondra, Cortidene, Dilar, Dillar, Flumethone, Haldrate, Haldrone, Metilar, Monocortin, Paramethasone 21-acetate, Paramethasone acetate, Paramezone, Stemex, Syntecort
IUPAC-namn [2-[(6S,8S,9S,10R,11S,13S,14S,16R,17R)-6-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
Specifikationer
Kemiskt namn eller material Paramethasone Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C24H31FO6/c1-12-7-16-15-9-18(25)17-8-14(27)5-6-22(17,3)21(15)19(28)10-23(16,4)24(12,30)20(29)11-31-13(2)26/h5-6,8,12,15-16,18-19,21,28,30H,7,9-11H2,1-4H3/t12-,15+,16+,18+,19+,21-,22+,23+,24+/m1/s1
Formel vikt 434.2105
Procent renhet >95%