Läs mer
Paraquat Monopyridone Iodide, TRC
Produktkod. 30425366
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425366 50 mg
30418492 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425366 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP19191050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35022-71-0
Molekylformel C12H13IN2O
Molekylvikt (g/mol) 328.15
Synonym 1',2'-Dihydro-1,1'-dimethyl-2'-oxo-4,4'-bipyridinium Iodide
IUPAC-namn 1,1'-dimethyl-2'-oxo-1',2'-dihydro-[4,4'-bipyridin]-1-ium iodide
LEDER O=C1C=C(C2=CC=[N+](C)C=C2)C=CN1C.[I-]
Specifikationer
Kemiskt namn eller material Paraquat Monopyridone Iodide
Kvantitet 50 mg
InChI formel InChI=1S/C12H13N2O.HI/c1-13-6-3-10(4-7-13)11-5-8-14(2)12(15)9-11;/h3-9H,1-2H3;1H/q+1;/p-1
Formel vikt 328.01