Läs mer
Tocris Bioscience™ Parathyroid hormone (1-34) (rat)
Parathyroid hormone (PTH) receptor agonist
Brand: Tocris Bioscience™ 6301/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Parathyroid hormone (1-34) (rat) is a parathyroid hormone (PTH) receptor agonist. Increases serum PTH levels and bone mass in rats.Specifikationer
| Rat | |
| 98614-76-7 | |
| Biological reactions | |
| Protein agonist | |
| RUO - research use only |
| 95% | |
| Store at −20°C | |
| 4057.74g/mol | |
| 1 mg | |
| AVSEIQLMHNLGKHLASVERMQWLRKKLQDVHNF |
Produkttitel
For Research Use Only