Läs mer
Pardoprunox Hydrochloride, TRC
Produktkod. 30410604
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30410604 5 mg
1 options
Produktkod. 30410604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2059085MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pardoprunox Hydrochloride
CAS 269718-83-4
Molekylformel C12 H15 N3 O2 . Cl H
Kvantitet 5 mg
Synonym Pardoprunox Hydrochloride, DU 126891 Hydrochloride, SLV 308, SLV 308 Hydrochloride, 7-(4-Methylpiperazin-1-yl)-3H-1,3-benzoxazol-2-one Hydrochloride
InChI formel InChI=1S/C12H15N3O2.ClH/c1-14-5-7-15(8-6-14)10-4-2-3-9-11(10)17-12(16)13-9;/h2-4H,5-8H2,1H3,(H,13,16);1H
LEDER Cl.CN1CCN(CC1)c2cccc3NC(=O)Oc23
IUPAC-namn 7-(4-methylpiperazin-1-yl)-3H-1,3-benzoxazol-2-one;hydrochloride
Molekylvikt (g/mol) 269.727
Formel vikt 269.093