missing translation for 'onlineSavingsMsg'
Learn More

PARG Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27808-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825845

  • 1656.73 SEK / 20µL
Estimated Shipment: 26-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.
Specifications

Specifications

PARG
Polyclonal
Unconjugated
PARG
PARG, PARG99
Rabbit
Antigen Affinity Chromatography
RUO
8505
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86W56
PARG
PARG Fusion Protein Ag27137
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers