Läs mer
Paricalcitol, TRC
Produktkod. 30010296
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010296 1 mg
30009357 25 mg
30005730 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1953001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 131918-61-1
Molekylformel C27 H44 O3
Molekylvikt (g/mol) 416.64
Synonym (1R,3R,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-Octahydro-1-[(1R,2E,4S)-5-hydroxy-1,4,5-trimethyl-2-hexen-1-yl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-1,3-cyclohexanediol, (1alpha,3beta,7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol, 19-Nor-1,25-
IUPAC-namn (1R,3R)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5S)-6-hydroxy-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]cyclohexane-1,3-diol
LEDER C[C@@]12[C@](\C(=C\C=C3C[C@@H](O)C[C@H](O)C3)\CCC1)(CC[C@@]2([C@@H](/C=C/[C@@H](C(C)(C)O)C)C)[H])[H]
Specifikationer
Kemiskt namn eller material Paricalcitol
Kvantitet 1 mg
InChI formel InChI=1S/C27H44O3/c1-18(8-9-19(2)26(3,4)30)24-12-13-25-21(7-6-14-27(24,25)5)11-10-20-15-22(28)17-23(29)16-20/h8-11,18-19,22-25,28-30H,6-7,12-17H2,1-5H3/b9-8+,21-11+/t18-,19+,22-,23-,24-,25+,27-/m1/s1
Formel vikt 416.329