Learn More
Draeger Safety™ Particle Filters
Clean breathable air from contaminants in a low-cost and effective manner. Dräger™ Particle Filters are suitable for all half and full face masks available on the market with connectors conforming to EN 148-1.
Brand: Draeger Safety™ 6732974
Additional Details : Weight : 0.01000kg
Description
- High level of user safety
- Individually packaged for optimum protection of the unused filter
- 12 year shelf life from date of production
- CE approved
Specifications
1140 P3 R | |
En 141:2000, EN 14387, EN 143:2000, EN 371, SAI-Global approved | |
Dräger X-plore™ Rd40 | |
Dräger X-plore™ 7500 |