missing translation for 'onlineSavingsMsg'
Learn More

PARK7/DJ-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11681-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878923

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
Specifications

Specifications

PARK7/DJ-1
0.55 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99497, Q99LX0
DJ 1, DJ1, Oncogene DJ1, PARK7, Parkinson disease protein 7, Protein DJ 1
Rabbit
IgG
20 μL
-20°C
Primary
11315, 57320
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Unconjugated
PARK7
Liquid
Park7
PARK7,DJ-1 Fusion Protein Ag2287
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
missing translation for 'documents'
Product Certifications
Ofertas especiales

Ofertas especiales