Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Run Tee
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand:  Cole-Parmer™ 31321-75
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 13407516
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
Compression Run Tee
Autoclavable
2 3/4 in.
1/4 in. OD X 1/4 in. NPT(M)
1/4 in.
Clear
Compression x NPT(M) Connection
PFA
1/4 in.
Autoclave or Ethylene Oxide
2 3/32 in.