Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Run Tee
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand: Cole-Parmer™ 31321-73
Additional Details : Weight : 0.02000kg
Description
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Compression Run Tee | |
Autoclavable | |
2 3/8 in. | |
1/8 in. OD X 1/4 in. NPT(M) | |
1/4 in. | |
Clear |
Compression x NPT(M) Connection | |
PFA | |
1/8 in. | |
Autoclave or Ethylene Oxide | |
1 9/16 in. |