Läs mer
Cole-Parmer™ Parker Hannifin™ PFA Compression Union Elbow
Slangstorlek:
1/2 tum
1/8 tum
Förpackningsstorlek:
Varje
Beskrivning
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifikationer
Specifikationer
| Beskrivning | Compression Connection |
| Autoklaveringsbar | Autoclavable |
| Material | PFA |
| Slangstorlek | 1/2 tum |
| Utloppsport | 1/2 x 1/2 in. |
| Steriliseringsmetod | Autoclave or Ethylene Oxide |
| Färg | Clear |
| Produkttyp | Compression Union Elbow |