Läs mer
Paromamine Trihydrochloride, TRC
Produktkod. 30136274
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136274 100 mg
30006187 10 mg
30012318 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136274 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP195340100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18685-97-7
Molekylformel C12 H25 N3 O7 . 3 H Cl
Molekylvikt (g/mol) 432.72
Synonym 4-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-2-deoxystreptamine Trihydrochloride;
IUPAC-namn (2R,3S,4R,5R,6S)-5-amino-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxy-cyclohexoxy]-2-(hydroxymethyl)tetrahydropyran-3,4-diol;trihydrochloride
LEDER Cl.Cl.Cl.N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2N)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Paromamine Trihydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H25N3O7.3ClH/c13-3-1-4(14)11(10(20)7(3)17)22-12-6(15)9(19)8(18)5(2-16)21-12;;;/h3-12,16-20H,1-2,13-15H2;3*1H/t3-,4+,5-,6-,7+,8-,9-,10-,11-,12-;;;/m1.../s1
Formel vikt 431.0993
Procent renhet >95%