missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PARP1 (Human) Recombinant Protein

Human PARP1 partial ORF ( AAH37545, 1 a.a. - 100 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000142-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181301

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MAESSDKLYRVEYAKSGRASCKKCSESIPKDSLRMAIMVQSPMFDGKVPHWYHFSCFWKVGHSIRHPDVEVDGFSELRWDDQQKVKKTAEAGGVTGKGQD
Specifications

Specifications

AAH37545
poly (ADP-ribose) polymerase 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PARP1
GST
142
Wheat germ expression system
10 μg
ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates