missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550719
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total PARP8 protein.
PARP8 Polyclonal antibody specifically detects PARP8 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
PARP8 | |
Polyclonal | |
Unconjugated | |
PARP8 | |
2810430O08Rik; ADP-ribosyltransferase diphtheria toxin-like 16; ARTD16; D13Ertd275e; I79_004209; Parp8; PARP-8; pART16; poly (ADP-ribose) polymerase family, member 8; poly [ADP-ribose] polymerase 8; poly(ADP-ribose) polymerase family member 8; Protein mono-ADP-ribosyltransferase PARP8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
52552, 79668 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q3UD82, Q8N3A8 | |
PARP8 | |
Synthetic peptide corresponding to residues near the C terminal of human poly (ADP-ribose) polymerase family, member 8 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |