Learn More
3M™ Particulate Filter, P2 R, 2128
3M™ Particulate Filters, 2128 connect to any half or full-face 3M™ Reusable Masks with a bayonet-style connection. Our P2 R filters are lightweight and small sized, with low breathing resistance.
Brand: 3M™ 7000052348
897.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
For breathing protection against solid and liquid particles, relief against nuisance-level organic vapours and acid gas, as well as protection against ozone up to 10 x OEL, use 3M™ Particulate Filters, 2128. They’re flexible enough to use across our range of reusable half and full-face masks, for safe working in hazardous environments. Our lightweight, P2 R filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and their small size means they fit comfortably under welding shields. They can be clicked into place quickly and easily using the 3M™ Bayonet Filter Attachment System, and they can be used with any 3M™ Reusable Masks with a bayonet-style connection.Specifications
EN143 : 2000, CE Approved | |
10 x TLV and organic vapours/acid gas below TLV, Solid and liquid particles. Plus ozone protection | |
P2R | |
Half- and Full-face masks |
Bayonet | |
White | |
Particulate Filter | |
1 Pair |