Learn More
Erlab™ Particulate Filter
Erlab™ Particulate Filter can be use with a range of filtrating fume hoods and storage cabinets.
Brand: Erlab™ I1019000001
Additional Details : Weight : 5.00000kg
Description
- Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.3 μm to optimize HEPA H14 filter life performance
- For use with powders
Specifications
HEPA H14 | |
CaptairFlex XLS 633 |