Learn More
CleanSpace Technology™ Particulate Filter P3 TM3 P SL R
The CleanSpace Particulate Filter P3 TM3 P SL R is suitable for protection against airborne particulates (dust, mists and fumes).
Brand:  CleanSpace Technology™ PAF-0035
Additional Details : Weight : 0.11550kg
Product Code. 15900062
Quantity Price
1 748.00 SEK / Each
10 672.15 SEK / Each
In stock, ready to ship: 5
In stock, ready to ship within 7 days: 30
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Suitable for protection against airborne particulate (dust, mists and fumes).
  • Materials: Fiberglass particulate media and plastic casing, silicone seal
  • Easy and quickly fitted and removed from the power unit
  • Pack of 3 (PAF-0035) or Pack of 24 (PAF-0059)
Specifications
Specifications
Filter
Videos
SDS
Documents
Special Offers
Special Offers
  • Use code 15003 to receive this promotion.
    : 15003
    2020-12-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/11/2020 and 31/12/2020 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More