Learn More
3M™ 6035 Model Particulate Filters
Offers P3 R protection against solid and liquid particles, hydrogen fluoride gas up to 30ppm and relief from nuisance level organic vapours and acid gases below nominal protection factor. The hard plastic case provides protection against sparks, heat and other harsh environments.
Brand:  3M™ 7000032083
Additional Details : Weight : 0.64000kg
Product Code. 15759007
  • 954.00 SEK / Pack of 10
Estimated Shipment
05-01-2021
Add to basket
Description
Description
  • Protective casing aids decontamination practices
  • Capable of providing protection against particulates at levels up to 10 times the Exposure Standard when used on a half-face respirator and up to 100 times the Expose Standard when used on a full-face respirator
  • Patented bonded web technology has more surface area than many standard filters which helps lower breathing resistance making it more comfortable to use
  • Unique case making it easy to perform a quick self-fit check
  • Temperature resistant cover protects filter media from heat, sparks or physical damage
  • 3M Bayonet Connection System ensures precise and secure locking
  • Can be used on all 3M 6000, 7000 and 7500 Series half and full respirators
  • Excellent for grinding, polishing, sanding, food manufacturing, construction/quarrying, woodworking and more
Specifications
Specifications
P3 R
Bayonet
Particulate Filter
Suitable for 3M™ 6000 and 3M™ 7000 Series Half Face and Full Face Masks
10/Pk.
EN143 : 2000
Protects against solids and liquid particles P3R, supplied with plastic cage for rough applications.
Grinding, sanding, polishing, food manufacturing, construction/quarrying, woodworking, pharmaceutical/powdered chemicals, water blasting, fiberglass installation, asbestos related tasks
6035