Läs mer
DiscoverX PathHunter™ CHO-K1 AGTRL1 Bioassay Kit
Beskrivning
The PathHunter™ AGTRL1 Bioassay kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This bioassay assesses ligand (e.g. apelin-13) based activation of AGTRL1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged AGTRL1 and EA-tagged β-Arrestin 2. Activation of AGTRL1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | β-Arrestin recruitment |
| För användning med (applikation) | Assay-Ready Bioassay Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | AGTRL1 |
| Celllinje | CHO-K1 |
| Mål | AGTRL1 |