Läs mer
DiscoverX PathHunter™ CHO-K1 GPR65 β-Arrestin Orphan GPCR Cell Line
Beskrivning
The PathHunter™ GPR65 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. protons) based activation of GPR65 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR65 and EA-tagged β-Arrestin 2. Activation of GPR65 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifikationer
Specifikationer
| Analysinträde | β-Arrestin recruitment |
| Celllinje | CHO-K1 |
| För användning med (applikation) | Cell Lines |
| Kvantitet | 2 injektionsflaskor |
| Målinträde | GPR65 |
| Tillträdesnummer | NM_003608 |