Läs mer
DiscoverX PathHunter™ eXpress ACVR1C/ACVR2B Dimerization Assay
Produktkod. 17394299
Encompass
Change view
Kvantitet:
200 dp (2 x 96-brunnar)
Produktkod. Kvantitet
17394299 200 dp (2 x 96-brunnar)
1 options
Produktkod. 17394299
Brand: DiscoverX 931047E3CP5M
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The PathHunter™ eXpress ACVR1C/ACVR2B Dimerization Assay measures ligand-induced dimerization of ACVR1C/ACVR2B . eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ ACVR1C/ACVR2B Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. activin B) induced dimerization of ACVR1C/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVR2B target protein fused to PK alongside ACVR1C target protein fused to EA. Upon ligand-induced dimerization of ACVR1C/ACVR2B , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Analysinträde ACVR1C/ACVR2B Dimerization
För användning med (applikation) Assay-Ready eXpress Kits
Primär agonist Activin B
Kvantitet 200 dp (2 x 96-brunnar)
Målinträde ACVR1C/ACVR2B
Celllinje U2OS
Mål ACVR1C/ACVR2B