Läs mer
DiscoverX PathHunter™ eXpress BMPR1A/BMPR2 Dimerization Assay
Beskrivning
The PathHunter™ BMPR1A/BMPR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. BMP-4) induced dimerization of BMPR1A/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR2 target protein fused to PK alongside BMPR1A target protein fused to EA. Upon ligand-induced dimerization of BMPR1A/BMPR2 , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | BMPR1A/BMPR2 Dimerization |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Primär agonist | BMP-4 |
| Kvantitet | 200 dp (2 x 96-brunnar) |
| Målinträde | BMPR1A/BMPR2 |
| Celllinje | U2OS |
| Mål | BMPR1A/BMPR2 |