Läs mer
DiscoverX PathHunter™ eXpress CALCR U2OS β-Arrestin GPCR Assay
Beskrivning
The PathHunter™ CALCR β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. calcitonin) based activation of CALCR GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged CALCR and EA-tagged β-Arrestin 2. Activation of CALCR stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | CALCR |
| Celllinje | U2OS |
| Mål | CALCR |