Läs mer
DiscoverX PathHunter™ eXpress CMKLR1 Activated GPCR Internalization Assay
Produktkod. 17393709
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
200 dp (2 x 96-brunnar)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17393709 200 dp (2 x 96-brunnar)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 17393709 Leverantör DiscoverX Leverantörsnummer 930720E3CP22M
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The PathHunter™ eXpress CMKLR1 Activated GPCR Internalization Assay measures CMKLR1 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ CMKLR1 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. chemerin) based internalization of CMKLR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CMKLR1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CMKLR1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Analysinträde GPCR
För användning med (applikation) Assay-Ready eXpress Kits
Kvantitet 200 dp (2 x 96-brunnar)
Målinträde CMKLR1
Mål CMKLR1