Läs mer
DiscoverX PathHunter™ eXpress EphB3 Functional Assay
Beskrivning
The PathHunter™ eXpress EphB3 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated EphB3 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged EphB3 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | Kinase Activity |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 200 dp (2 x 96-brunnar) |
| Målinträde | EphB3 |
| Mål | EphB3 |