Läs mer
DiscoverX PathHunter™ eXpress GLP1R CHO-K1 β-Arrestin-1 GPCR Assay
Beskrivning
The PathHunter™ GLP1R (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. exendin-4) based activation of GLP1R GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GLP1R and EA-tagged β-Arrestin-1. Activation of GLP1R stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | β-arrestin recruitment |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Primär agonist | Exendin-4 |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | GLP1R (ARR1) |
| Celllinje | CHO-K1 |
| Mål | GLP1R (ARR1) |