Learn More
DiscoverX PathHunter™ eXpress GPR132 CHO-K1 β-Arrestin Orphan GPCR Assay
Description
The PathHunter™ GPR132 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR132 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR132 and EA-tagged β-Arrestin 2. Activation of GPR132 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifications
Specifications
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | GPR132 |
| Celllinje | CHO-K1 |
| Mål | GPR132 |