Learn More
DiscoverX PathHunter™ eXpress GPR132 CHO-K1 β-Arrestin Orphan GPCR Assay
Product Code. 17393139
Encompass
Change view
Click to view available options
Kvantitet:
1 000 dp (10 x 96-brunnar)
Product Code. Kvantitet
17393139 1 000 dp (10 x 96-brunnar)
1 options
Product Code. 17393139 Supplier DiscoverX Supplier No. 930411E2ACP1L
 Begränsad artikel
Beräknad leverans: 24-09-2026
Add to basket
Add to basket
The PathHunter™ eXpress GPR132 CHO-K1 β-Arrestin Orphan GPCR Assay measures GPR132 (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ GPR132 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR132 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR132 and EA-tagged β-Arrestin 2. Activation of GPR132 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifications
Analysinträde GPCR
För användning med (applikation) Assay-Ready eXpress Kits
Kvantitet 1 000 dp (10 x 96-brunnar)
Målinträde GPR132
Celllinje CHO-K1
Mål GPR132