Läs mer
DiscoverX PathHunter™ eXpress GPR27 CHO-K1 β-Arrestin Orphan GPCR Assay
Produktkod. 17382999
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 000 dp (10 x 96-brunnar)
Produktkod. Kvantitet
17382999 1 000 dp (10 x 96-brunnar)
1 options
Produktkod. 17382999
Brand: DiscoverX 930334E2ACP1L
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The PathHunter™ eXpress GPR27 CHO-K1 β-Arrestin Orphan GPCR Assay measures GPR27 (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ GPR27 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR27 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR27 and EA-tagged β-Arrestin 2. Activation of GPR27 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifikationer
Analysinträde GPCR
För användning med (applikation) Assay-Ready eXpress Kits
Formatera 1,000 dp (10 x 96-well)
Kvantitet 1 000 dp (10 x 96-brunnar)
Målinträde GPR27
Celllinje CHO-K1
Mål GPR27