Läs mer
DiscoverX PathHunter™ eXpress HCRTR1 Total GPCR Internalization Assay
Beskrivning
The PathHunter™ HCRTR1 Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. orexin A) based internalization of HCRTR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged HCRTR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the HCRTR1 receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | HCRTR1 |
| Mål | HCRTR1 |