Läs mer
DiscoverX PathHunter™ eXpress OPRD1 Activated GPCR Internalization Assay
Beskrivning
The PathHunter™ OPRD1 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. DADLE/DPDPE) based internalization of OPRD1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged OPRD1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting OPRD1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 200 dp (2 x 96-brunnar) |
| Målinträde | OPRD1 |
| Mål | OPRD1 |