Läs mer
DiscoverX PathHunter™ eXpress OPRD1 Activated GPCR Internalization Assay
Produktkod. 17363629
Encompass
Change view
Kvantitet:
200 dp (2 x 96-brunnar)
Produktkod. Kvantitet
17363629 200 dp (2 x 96-brunnar)
1 options
Produktkod. 17363629 Leverantör DiscoverX Leverantörsnummer 930673E3CP0M
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The PathHunter™ eXpress OPRD1 Activated GPCR Internalization Assay measures OPRD1 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ OPRD1 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. DADLE/DPDPE) based internalization of OPRD1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged OPRD1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting OPRD1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Analysinträde GPCR
För användning med (applikation) Assay-Ready eXpress Kits
Kvantitet 200 dp (2 x 96-brunnar)
Målinträde OPRD1
Mål OPRD1