Läs mer
DiscoverX PathHunter™ eXpress P2RY2 U2OS β-Arrestin GPCR Assay
Beskrivning
The PathHunter™ P2RY2 β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. UTP) based activation of P2RY2 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged P2RY2 and EA-tagged β-Arrestin 2. Activation of P2RY2 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | P2RY2 |
| Celllinje | U2OS |
| Mål | P2RY2 |