Läs mer
DiscoverX PathHunter™ eXpress rGPR35 CHO-K1 β-Arrestin GPCR Assay
Beskrivning
The PathHunter™ rGPR35 β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. zaprinast) based activation of rGPR35 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged rGPR35 and EA-tagged β-Arrestin 2. Activation of rGPR35 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein.
Specifikationer
Specifikationer
| Analysinträde | β-Arrestin-2 recruitment |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Primär agonist | Zaprinast |
| Kvantitet | 200 dp (2 x 96-brunnar) |
| Målinträde | rGPR35 |
| Celllinje | CHO-K1 |
| Mål | rGPR35 |