Läs mer
DiscoverX PathHunter™ eXpress SSTR2 Activated GPCR Internalization Assay
Produktkod. 17393629
Encompass
Change view
Kvantitet:
1 000 dp (10 x 96-brunnar)
Produktkod. Kvantitet
17393629 1 000 dp (10 x 96-brunnar)
1 options
Produktkod. 17393629
Brand: DiscoverX 930676E2CP0L
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The PathHunter™ eXpress SSTR2 Activated GPCR Internalization Assay measures SSTR2 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ SSTR2 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. somatostatin 28) based internalization of SSTR2 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged SSTR2. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting SSTR2-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Analysinträde GPCR
För användning med (applikation) Assay-Ready eXpress Kits
Kvantitet 1 000 dp (10 x 96-brunnar)
Målinträde SSTR2
Mål SSTR2