Läs mer
DiscoverX PathHunter™ eXpress SSTR2 Activated GPCR Internalization Assay
Beskrivning
The PathHunter™ SSTR2 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. somatostatin 28) based internalization of SSTR2 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged SSTR2. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting SSTR2-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifikationer
Specifikationer
| Analysinträde | GPCR |
| För användning med (applikation) | Assay-Ready eXpress Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | SSTR2 |
| Mål | SSTR2 |