Läs mer
DiscoverX PathHunter™ eXpress TrkA Functional Assay
Produktkod. 17343209
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
200 dp (2 x 96-brunnar)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17343209 200 dp (2 x 96-brunnar)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 17343209 Leverantör DiscoverX Leverantörsnummer 930462E3CP16M
The PathHunter™ eXpress TrkA Functional Assay measures TrkA receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ eXpress TrkA Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkA dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged TrkA and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifica
Analysinträde Kinase
För användning med (applikation) Assay-Ready eXpress Kits
Kvantitet 200 dp (2 x 96-brunnar)
Målinträde TrkA
Mål TrkA
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.