Läs mer
DiscoverX PathHunter™ Tocilizumab Bioassay Kit
Description
The PathHunter™ Tocilizumab Bioassay kit provides an easy-to-use cell based assay to determine potency of anti-IL-6R or anti-IL-6 antibodies such as Tocilizumab. This bioassay assesses ligand (e.g. IL-6) induced dimerization of of IL-6ST and IL-6R. receptors. This product uses EFC technology. The cell line included overexpresses IL-6ST target protein fused to PK alongside IL-6R target protein fused to EA. Upon ligand-induced dimerization of IL-6ST and IL-6R , detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Analysinträde | Dimerization |
| För användning med (applikation) | Assay-Ready Bioassay Kits |
| Kvantitet | 1 000 dp (10 x 96-brunnar) |
| Målinträde | IL-6R/IL-6ST |
| Celllinje | U2OS |
| Mål | IL-6R/IL-6ST |