Läs mer
Intelsius™ PathoSorb Absorbent Pouch
Produktkod. 17328589
Encompass_Preferred
Change view
Kapacitet:
152 ml
215 ml
Mått (L x B):
225 x 125 mm (8,9 x 4,9 tum)
325 x 125 mm (12,8 x 4,9 tum)
Inkluderar:
4 fack
6 fack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kapacitet Mått (L x B) Inkluderar
17328589 215 ml 325 x 125 mm (12,8 x 4,9 tum) 6 fack
17308589 152 ml 225 x 125 mm (8,9 x 4,9 tum) 4 fack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17328589 Leverantör Intelsius™ Leverantörsnummer AB008
Beräknad leverans: 10-11-2026
Protect and separate sample vials with this PathoSorb Absorbent sample Pouch.
  • Keeps sample tubes safe and secure
  • Lint free
  • Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations
Specifikationer
Inkluderar 6 fack
Certifikat och efterlevnad UN3373, UN2900, UN2814
Typ Absorbent Sample Pouch
Mått (L x B) 325 x 125 mm (12,8 x 4,9 tum)
Kapacitet 215 ml