Läs mer
PBDE 203, TRC
Produktkod. 17949309
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17949309 10 mg
17947919 25 mg
17917999 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17949309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP21523010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 337513-72-1
Molekylformel C12 H2 Br8 O
Molekylvikt (g/mol) 801.38
Synonym Benzene, 1,2,3,4,5-pentabromo-6-(2,4,5-tribromophenoxy)-, Benzene, pentabromo(2,4,5-tribromophenoxy)- (9CI), 1,2,3,4,5-Pentabromo-6-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5,5',6-Octabromodiphenyl ether, BDE 203, PBDE 203
IUPAC-namn 1,2,3,4,5-pentabromo-6-(2,4,5-tribromophenoxy)benzene
LEDER Brc1cc(Br)c(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)cc1Br
Specifikationer
Kemiskt namn eller material PBDE 203 (2,2',3,4,4',5,5',6-Octabromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H2Br8O/c13-3-1-5(15)6(2-4(3)14)21-12-10(19)8(17)7(16)9(18)11(12)20/h1-2H
Formel vikt 793.3573
Procent renhet >95%