missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ PBOV1 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 18015521
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18015521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18015521 Leverantör Invitrogen™ Leverantörsnummer PA5144975

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: YSIEQSHHAI LTPLQTHLTM KGSSMKCSSL SSEAILFT Antigen sequence identity: Mouse 38%; Rat 33%.

This intronless gene encodes a protein of unknown function. Its expression is up-regulated in some types of cancer, including prostate, breast, and bladder cancer. [provided by RefSeq, Aug 2011].
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen PBOV1
Användningsområden Immunocytochemistry
Klassificering Polyclonal
Koncentration 0.05 mg/mL
Konjugera Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen PBOV1
Genaccessionsnr. Q9GZY1
Gene Alias dJ171N11.2; PBOV1; prostate and breast cancer overexpressed 1; prostate and breast cancer overexpressed gene 1 protein; Protein UROC28; UC28; UROC28
Gensymboler PBOV1
Värd art Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 71-108. Recombinant protein control fragment (Product #RP-109812).
Reningsmetod Affinity Chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 59351
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Formatera Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.