missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PBP Recombinant Protein
4515.00 SEK - 6845.00 SEK
Specifications
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Molecular Weight (g/mol) | 46.31 |
pH Range | 8 |
Preparation Method | In vitro wheat germ expression system |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178651
|
Abnova™
H00005037-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16188651
|
Abnova™
H00005037-P01.25UG |
25μg |
6845.00 SEK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, Protein Array, ELISA, Western Blot | |
46.31 | |
In vitro wheat germ expression system | |
Wheat Germ (in vitro) | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Human | |
Solution |