missing translation for 'onlineSavingsMsg'
Learn More

PBRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12563-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840334

  • 4721.67 SEK / 150µL
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This locus encodes a subunit of ATP-dependent chromatin-remodeling complexes. The encoded protein has been identified as in integral component of complexes necessary for ligand-dependent transcriptional activation by nuclear hormone receptors. Mutations at this locus have been associated with primary clear cell renal cell carcinoma.
Specifications

Specifications

PBRM1
Polyclonal
Unconjugated
PBRM1
BAF180, BRG1 associated factor 180, hPB1, PB1, PBRM1, polybromo 1, Polybromo 1D, Protein polybromo 1
Rabbit
Antigen Affinity Chromatography
RUO
306254, 55193, 66923
-20°C
Liquid
Immunoprecipitation, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86U86, Q8BSQ9
PBRM1
PBRM1 Fusion Protein Ag3148
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers