missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PCP4 (Human) Recombinant Protein

Human PCP4 full-length ORF ( AAH13791, 1 a.a. - 62 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005121-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118811

  • 6150.00 SEK / 25µg
Estimated Shipment: 13-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSERQGAGATNGKDKTSGENDGQKKVQEEFDIDMDAPETERAAVAIQSQFRKFQKKKAGSQS
Specifications

Specifications

AAH13791
Purkinje cell protein 4
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCP4
GST
5121
Wheat germ expression system
25 μg
PEP-19
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers