Learn More
PCR WaterAbnova™
Product Code. 16072565
Change view
Click to view available options
Quantity:
10 x 1.8 mL
25 x 1.8 mL
Unit Size:
Pack of 10
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16072565 25 x 1.8 mL Pack of 25
16062565 10 x 1.8 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16072565 Supplier Abnova Supplier No. D0022.25x1.8mL
 Restricted Item
Beräknad leverans: 15-01-2027
Manufactured and tested for use as a component in PCR reactions, Validated as well for use with highly sensitive PCR reaction including those with eubacterial primer sets.
Stored in convenient 2 mL tubes to avoid unnecessary melting and freezing of the whole water supply.
Specifications
Quantity 25 x 1.8 mL