Learn More
Hamilton™ Union 1/4-28 Threads
An economical solution for making connections in the laboratory.
Brand: Hamilton™ 88800
Additional Details : Weight : 0.01000kg
Description
- Manufactured from CTFE
- Sold in packs of 10
Specifications
PCTFE | |
Coupling |