Läs mer
Tocris Bioscience™ PD 0332991 isethionate
GREENER_CHOICE
Produktkod. 30001694
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001694 50 mg
30001693 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001694 Leverantör Tocris Bioscience™ Leverantörsnummer 4786/50
Potent, selective Cdk4/6 inhibitor; brain penetrant
Potent cyclin-dependent kinase 4 (cdk4) and cdk6 inhibitor (IC50 values are 11 nM and 15 nM respectively). Also displays activity at cdk2, cdk5 and cdk9 (IC50 values reported to be 1.6, 1.8 and 0.42-1.1 μM, respectively). Induces G1 cell cycle arrest and senescence in retinoblastoma protein (Rb)-proficient cell lines; blocks growth of intracranial glioblastoma multiforme xenografts in mice. Brain penetrant.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 827022-33-3
Molekylformel C26H35N7O6S
Specifikationer
Kemiskt namn eller material PD 0332991 isethionate
Analysprocentintervall >98%
Kvantitet 50 mg
Formel vikt 573.66
Molekylvikt (g/mol) 573.669