Läs mer
Tocris Bioscience™ PD 102807
GREENER_CHOICE
Produktkod. 18005042
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18005042 10 mg
18045012 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18005042 Leverantör Tocris Bioscience™ Leverantörsnummer 1671/10
Selective M4 antagonist
Selective M4 muscarinic receptor antagonist. IC50 values are 91, 6559, 3441, 950 and 7412 nM for human M4, M1, M2, M3, and M5 receptors respectively.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23062-91-1
Molekylformel C23H24N2O4
Specifikationer
Kemiskt namn eller material PD 102807
Kvantitet 10 mg
Formel vikt 392.45
Renhet >98%
Molekylvikt (g/mol) 392.455