Läs mer
Tocris Bioscience™ PD 146176
GREENER_CHOICE
Produktkod. 30001341
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001341 10 mg
30002119 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001341 Leverantör Tocris Bioscience™ Leverantörsnummer 2850/10
Selective 15-lipoxygenase inhibitor
Specific, non-competitve 15-lipoxygenase (15-LOX) inhibitor (Ki = 197nM) that has no demonstrable effect on 5-LOX, 12-LOX, COX-1 or COX-2 (IC50 = 0.54 μM for 15-LOX in rabbit reticulocytes). Lacks non-specific antioxidant properties and prevents atherogenesis via regulation of monocyte-macrophage enrichment in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4079-26-9
Molekylformel C15H11NS
Specifikationer
Kemiskt namn eller material PD 146176
Kvantitet 10 mg
Formel vikt 237.32
Renhet >98%
Molekylvikt (g/mol) 237.32